Benzo[b]furan-3-boronic acid, pinacol ester 98%

Supplier: Ambeed
Warning

Synonyms: 3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-1-benzofuran, 2-(Benzofuran-3-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-(1-Benzofuran-3-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

A680346-1G A680346-5G A680346-250MG
76879-522EA 164.44 USD
76879-522 76879-524 77614-098
Benzo[b]furan-3-boronic acid, pinacol ester 98%
Benzo[b]furan-3-boronic acid, pinacol ester

Precaución: for Research Use Only

Formula: C₁₄H₁₇BO₃
MW: 244.094 g/mol
MDL Number: MFCD11841067
CAS Number: 796851-30-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR