4-Amino-1,3-dihydro-2H-benzimidazol-2-one 98%

Supplier: Ambeed
Warning

A677908-100MG A677908-1G A677908-250MG
76879-504EA 84.01 USD
76879-504 76879-506 76879-508
4-Amino-1,3-dihydro-2H-benzimidazol-2-one 98%
4-Amino-1,3-dihydro-2H-benzimidazol-2-one

Precaución: for Research Use Only

Formula: C₇H₇N₃O
MDL Number: MFCD18089179
CAS Number: 75370-65-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR