5-(tert-Butyl)-2-propoxybenzene-1-sulfonyl chloride 95+%

Supplier: Ambeed
Danger

A676279-100MG A676279-1G A676279-250MG
76879-492EA 52 USD
76879-492 76879-494 76879-496
5-(tert-Butyl)-2-propoxybenzene-1-sulfonyl chloride 95+%
5-(tert-Butyl)-2-propoxybenzene-1-sulfonyl chloride

Precaución: for Research Use Only

Formula: C₁₃H₁₉ClO₃S
MW: 290.81 g/mol
MDL Number: MFCD03789028
CAS Number: 681260-21-9
UN: 3261
ADR: 8 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR