Methyl cis-2-aminocyclopentanecarboxylate hydrochloride 98%

Supplier: Ambeed
Warning

A676092-100MG A676092-1G A676092-250MG A676092-5G
76879-486EA 25 USD
76879-486 76879-488 76879-490 77884-570
Methyl cis-2-aminocyclopentanecarboxylate hydrochloride 98%
Methyl cis-2-aminocyclopentanecarboxylate hydrochloride

Precaución: for Research Use Only

Formula: C₇H₁₃NO₂·ClH
MDL Number: MFCD09997947
CAS Number: 119993-55-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR