4-Fluoro-2-n-propoxybenzeneboronic acid 95+%

Supplier: Ambeed
Warning

A673883-100MG A673883-250MG A673883-5G A673883-1G
76879-478EA 23 USD
76879-478 76879-480 76879-482 77886-596
4-Fluoro-2-n-propoxybenzeneboronic acid 95+%
4-Fluoro-2-n-propoxybenzeneboronic acid

Precaución: for Research Use Only

Formula: C₉H₁₂BFO₃
MDL Number: MFCD06411288
CAS Number: 480438-60-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR