2-Methyl-2-phenylpropan-1-ol 97%

Supplier: Ambeed
Warning

Synonyms: 2-Methyl-2-phenyl-1-propanol

A673770-100MG A673770-1G A673770-250MG A673770-5G
76879-472EA 41.38 USD
76879-472 76879-474 76879-476 77673-842
2-Methyl-2-phenylpropan-1-ol 97%
2-Methyl-2-phenylpropan-1-ol

Precaución: for Research Use Only

Formula: C₁₀H₁₄O
MW: 150.218 g/mol
MDL Number: MFCD00506139
CAS Number: 2173-69-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR