4-Benzylcarbamoyl-3-fluorophenylboronic acid 97%

Supplier: Ambeed
Warning

Synonyms: (4-(Benzylcarbamoyl)-3-fluorophenyl)boronic acid, 4-(Benzylcarbamoyl)-3-fluorophenylboronic acid

A668508-1G A668508-250MG A668508-5G
76879-452EA 166.57 USD
76879-452 76879-454 76879-456
4-Benzylcarbamoyl-3-fluorophenylboronic acid 97%
4-Benzylcarbamoyl-3-fluorophenylboronic acid

Precaución: for Research Use Only

Formula: C₁₄H₁₃BFNO₃
MW: 273.0700 g/mol
MDL Number: MFCD08689501
CAS Number: 874289-19-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR