2,2'-Bipyridine-5,5'-dicarbonitrile 97%

Supplier: Ambeed
Warning

Synonyms: [2,2'-Bipyridine]-5,5'-dicarbonitrile

A665739-100MG A665739-250MG
76879-442EA 45 USD
76879-442 76879-444
2,2'-Bipyridine-5,5'-dicarbonitrile 97%
2,2'-Bipyridine-5,5'-dicarbonitrile

Precaución: for Research Use Only

Formula: C₁₂H₆N₄
MW: 206.2028 g/mol
MDL Number: MFCD11855995
CAS Number: 1802-29-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR