4,5-Dichloro-o-toluidine 98%

Supplier: Ambeed
Warning

A665416-1G A665416-250MG
76879-438EA 39 USD
76879-438 76879-440
4,5-Dichloro-o-toluidine 98%
4,5-Dichloro-o-toluidine

Precaución: for Research Use Only

Formula: C₇H₇Cl₂N
MDL Number: MFCD11855825
CAS Number: 2387-08-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR