Ethyl (R)-4-phenyl-2-(trifluoromethanesulfonyloxy)butyrate 95%

Supplier: Ambeed
Warning

Synonyms: Ethyl (R)-4-phenyl-2-[[(trifluoromethyl)sulphonyl]oxy]butyrate, (R)-4-Phenyl-2-(trifluoromethanesulphonyloxy)butyric acid Ethyl Ester

A664290-1G A664290-250MG A664290-5G
76879-424EA 33 USD
76879-424 76879-426 76879-428
Ethyl (R)-4-phenyl-2-(trifluoromethanesulfonyloxy)butyrate 95%
Ethyl (R)-4-phenyl-2-(trifluoromethanesulfonyloxy)butyrate

Precaución: for Research Use Only

Formula: C₁₃H₁₅F₃O₅S
MW: 340.31 g/mol
MDL Number: MFCD09751096
CAS Number: 88767-98-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR