4-Isopropylbenzene-1,3-diol 97%

Supplier: Ambeed
Warning

Synonyms: 1,3-Dihydroxy-4-(prop-2-yl)benzene, 4-Isopropylresorcinol

A663542-5G A663542-1G
76879-422EA 37.14 USD
76879-422 77673-816
4-Isopropylbenzene-1,3-diol 97%
4-Isopropylbenzene-1,3-diol

Precaución: for Research Use Only

Formula: C₉H₁₂O₂
MW: 152.19 g/mol
MDL Number: MFCD05663778
CAS Number: 23504-03-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR