Ethyl 4-oxotetrahydrothiophene-3-carboxylate 98%

Supplier: Ambeed
Warning

A662993-100MG A662993-1G A662993-250MG A662993-5G
76879-414EA 31.83 USD
76879-414 76879-416 76879-418 76879-420
Ethyl 4-oxotetrahydrothiophene-3-carboxylate 98%
Ethyl 4-oxotetrahydrothiophene-3-carboxylate

Precaución: for Research Use Only

Formula: C₇H₁₀O₃S
MW: 174.218 g/mol
MDL Number: MFCD01318533
CAS Number: 78647-31-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR