Hydroxy(1,5-cyclooctadiene)rhodium(I) dimer 96%

Supplier: Ambeed
Warning

A662257-1G A662257-250MG A662257-100MG
76879-410EA 560 USD
76879-410 76879-412 77569-260
Hydroxy(1,5-cyclooctadiene)rhodium(I) dimer 96%
Hydroxy(1,5-cyclooctadiene)rhodium(I) dimer

Precaución: For research use only.

Formula: C₁₆H₂₆O₂Rh₂
MDL Number: MFCD04974244
CAS Number: 73468-85-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR