6,7-Dichloroquinoxaline 95%

Supplier: Ambeed
Warning

A660431-1G A660431-250MG A660431-5G
76879-388EA 66 USD
76879-388 76879-390 76879-392
6,7-Dichloroquinoxaline 95%
6,7-Dichloroquinoxaline

Precaución: for Research Use Only

Formula: C₈H₄Cl₂N₂
MW: 199.0368 g/mol
MDL Number: MFCD00456885
CAS Number: 19853-64-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR