2-(Methylamino)-5-pyrimidineboronic acid 98%

Supplier: Ambeed
Warning

Synonyms: (2-(Methylamino)pyrimidin-5-yl)boronic acid

A658451-100MG A658451-1G A658451-250MG A658451-5G
76879-374EA 36.08 USD
76879-374 76879-376 76879-378 76879-380
2-(Methylamino)-5-pyrimidineboronic acid 98%
2-(Methylamino)-5-pyrimidineboronic acid

Precaución: for Research Use Only

Formula: C₅H₈BN₃O₂
MW: 152.9500 g/mol
MDL Number: MFCD16618931
CAS Number: 1033745-26-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR