Dibutyl [2,2'-bipyridine]-4,4'-dicarboxylate 98%

Supplier: Ambeed
Warning

A657766-100MG A657766-1G A657766-250MG
76879-366EA 106.09 USD
76879-366 76879-368 76879-370
Dibutyl [2,2'-bipyridine]-4,4'-dicarboxylate 98%
Dibutyl [2,2'-bipyridine]-4,4'-dicarboxylate

Precaución: for Research Use Only

Formula: C₂₀H₂₄N₂O₄
MW: 356.41 g/mol
MDL Number: MFCD11042474
CAS Number: 69641-93-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR