2-(tert-Butyl)-4,5-dichloro-3(2H)-pyridazinone 95%

Supplier: Ambeed
Warning

Synonyms: 2-(tert-Butyl)-4,5-dichloropyridazin-3(2H)-one

A655552-1G A655552-250MG A655552-5G
76879-358EA 28.65 USD
76879-358 76879-360 76879-362
2-(tert-Butyl)-4,5-dichloro-3(2H)-pyridazinone 95%
2-(tert-Butyl)-4,5-dichloro-3(2H)-pyridazinone

Precaución: for Research Use Only

Formula: C₈H₁₀Cl₂N₂O
MW: 221.09 g/mol
MDL Number: MFCD00277876
CAS Number: 84956-71-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR