2-Acetamido-4-(trifluoromethyl)benzeneboronic acid, pinacol ester 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
N-[2-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-5-(trifluoromethyl)phenyl]acetamide, 3-(Acetylamino)-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzotrifluoride, N-(2-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-5-(trifluoromethyl)phenyl)acetamide
A652659-100MG
A652659-1G
A652659-250MG
A652659-5G
76879-344EA
36
USD
76879-344
76879-346
76879-348
76879-350
2-Acetamido-4-(trifluoromethyl)benzeneboronic acid, pinacol ester 98%
2-Acetamido-4-(trifluoromethyl)benzeneboronic acid, pinacol ester
Precaución: for Research Use Only
Formula:
C₁₅H₁₉BF₃NO₃ |
MDL Number:
MFCD12026072 CAS Number: 1150271-57-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...