3,4-Bis(methoxycarbonyl)phenylboronic acid 98%

Supplier: Ambeed
Warning

Synonyms: (3,4-Bis(methoxycarbonyl)phenyl)boronic acid

A651807-100MG A651807-1G A651807-250MG
76879-340EA 30 USD
76879-340 76879-342 77884-522
3,4-Bis(methoxycarbonyl)phenylboronic acid 98%
3,4-Bis(methoxycarbonyl)phenylboronic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₁BO₆
MW: 238.0017 g/mol
MDL Number: MFCD11504854
CAS Number: 1072951-51-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR