2-((S)-3-(((S)-1-Ethoxy-1-oxo-4-phenylbutan-2-yl)amino)-2-oxo-2,3,4,5-tetrahydro-1H-benzo[b]azepin-1-yl)acetic acid 98%

Supplier: Ambeed
Warning

A651804-1MG A651804-250MG A651804-100MG
76879-336EA 6 USD
76879-336 76879-338 77884-521
2-((S)-3-(((S)-1-Ethoxy-1-oxo-4-phenylbutan-2-yl)amino)-2-oxo-2,3,4,5-tetrahydro-1H-benzo[b]azepin-1-yl)acetic acid 98%
2-((S)-3-(((S)-1-Ethoxy-1-oxo-4-phenylbutan-2-yl)amino)-2-oxo-2,3,4,5-tetrahydro-1H-benzo[b]azepin-1-yl)acetic acid

Precaución: for Research Use Only

Formula: C₂₄H₂₈N₂O₅
MDL Number: MFCD00864466
CAS Number: 86541-75-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR