5'-Bromospiro[cyclopropane-1,3'-indolin]-2'-one 98%

Supplier: Ambeed
Warning

A650140-100MG A650140-1G A650140-250MG
76879-330EA 48.81 USD
76879-330 76879-332 76879-334
5'-Bromospiro[cyclopropane-1,3'-indolin]-2'-one 98%
5'-Bromospiro[cyclopropane-1,3'-indolin]-2'-one

Precaución: for Research Use Only

Formula: C₁₀H₈BrNO
MW: 238.08 g/mol
MDL Number: MFCD13195328
CAS Number: 875071-97-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR