Ethyl 2,2-dichloro-3-oxobutanoate 95%

Supplier: Ambeed
Danger

A645219-1G A645219-250MG A645219-25G A645219-5G
76879-284EA 100.79 USD
76879-284 76879-286 76879-288 76879-290
Ethyl 2,2-dichloro-3-oxobutanoate 95%
Ethyl 2,2-dichloro-3-oxobutanoate

Precaución: for Research Use Only

Formula: C₆H₈Cl₂O₃
MW: 199.032 g/mol
MDL Number: MFCD00129136
CAS Number: 6134-66-3
UN: 2920
ADR: 8,3,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR