3-Bromo-5-fluoro-2-hydroxybenzaldehyde 95%

Supplier: Ambeed
Warning

A643499-1G A643499-250MG A643499-5G A643499-25G
76879-278EA 20 USD
76879-278 76879-280 76879-282 77884-505
3-Bromo-5-fluoro-2-hydroxybenzaldehyde 95%
3-Bromo-5-fluoro-2-hydroxybenzaldehyde

Precaución: for Research Use Only

Formula: C₇H₄BrFO₂
MDL Number: MFCD08458073
CAS Number: 178546-34-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR