3-(2-Ethoxy-2-oxoethoxy)benzeneboronic acid 98%

Supplier: Ambeed
Warning

Synonyms: (3-(2-Ethoxy-2-oxoethoxy)phenyl)boronic acid

A643490-1G A643490-250MG A643490-5G
76879-272EA 102 USD
76879-272 76879-274 76879-276
3-(2-Ethoxy-2-oxoethoxy)benzeneboronic acid 98%
3-(2-Ethoxy-2-oxoethoxy)benzeneboronic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₃BO₅
MDL Number: MFCD09878321
CAS Number: 957062-63-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR