1-Methylindole-3-boronic acid pinacol ester 96%
Supplier: Ambeed
|
Warning
|
Synonyms:
1-Methyl-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indole, N-Methylindole-3-boronic acid pinacol ester
A642300-100MG
A642300-1G
A642300-250MG
A642300-25G
A642300-5G
A642300-10G
76879-262EA
13
USD
76879-262
76879-264
76879-266
76879-268
76879-270
77884-503
1-Methylindole-3-boronic acid pinacol ester 96%
1-Methylindole-3-boronic acid pinacol ester
Precaución: for Research Use Only
Formula:
C₁₅H₂₀BNO₂ MW: 257.1358 g/mol |
MDL Number:
MFCD11894338 CAS Number: 683229-61-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...