4,4,5,5-Tetramethyl-2-[4-[(tetrahydro-2H-pyran-2-yl)oxy]phenyl]-1,3,2-dioxaborolane 97%

Supplier: Ambeed
Warning

Synonyms: 2-[4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolane-2-yl)phenoxy]tetrahydro-2H-pyran, 4,4,5,5-Tetramethyl-2-(4-((tetrahydro-2H-pyran-2-yl)oxy)phenyl)-1,3,2-dioxaborolane, 4-[(Tetrahydro-2H-pyran-2-yl)oxy]phenylboronic acid Pinacol ester

A638779-10G A638779-1G A638779-250MG A638779-5G
76879-246EA 101.85 USD
76879-246 76879-248 76879-250 76879-252
4,4,5,5-Tetramethyl-2-[4-[(tetrahydro-2H-pyran-2-yl)oxy]phenyl]-1,3,2-dioxaborolane 97%
4,4,5,5-Tetramethyl-2-[4-[(tetrahydro-2H-pyran-2-yl)oxy]phenyl]-1,3,2-dioxaborolane

Precaución: for Research Use Only

Formula: C₁₇H₂₅BO₄
MW: 304.19 g/mol
MDL Number: MFCD06659834
CAS Number: 889865-38-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR