2,6-Dibromo-4-nitroaniline 98%

Supplier: Ambeed
Warning

A638442-1000G A638442-100G A638442-500G
76879-240EA 83.82 USD
76879-240 76879-242 76879-244
2,6-Dibromo-4-nitroaniline 98%
2,6-Dibromo-4-nitroaniline

Precaución: for Research Use Only

Formula: C₆H₄Br₂N₂O₂
MW: 295.92 g/mol
MDL Number: MFCD00007639
CAS Number: 827-94-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR