5-(Trifluoromethoxy)picolinonitrile 95%

Supplier: Ambeed
Warning

A636705-100MG A636705-250MG
76879-236EA 209 USD
76879-236 76879-238
5-(Trifluoromethoxy)picolinonitrile 95%
5-(Trifluoromethoxy)picolinonitrile

Precaución: for Research Use Only

Formula: C₇H₃F₃N₂O
MW: 188.10 g/mol
MDL Number: MFCD17169840
CAS Number: 1206978-22-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR