1-(2-Amino-4-(trifluoromethyl)phenyl)ethanone 98%

Supplier: Ambeed
Warning

A635794-100MG A635794-1G A635794-250MG A635794-5G
76879-228EA 32.89 USD
76879-228 76879-230 76879-232 76879-234
1-(2-Amino-4-(trifluoromethyl)phenyl)ethanone 98%
1-(2-Amino-4-(trifluoromethyl)phenyl)ethanone

Precaución: for Research Use Only

Formula: C₉H₈F₃NO
MW: 203.16 g/mol
MDL Number: MFCD11113374
CAS Number: 37885-07-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR