6-(Hydroxymethyl)pyridine-2-carboxaldehyde 98%

Supplier: Ambeed
Danger

A632955-1G A632955-100MG A632955-250MG
76879-210EA 265 USD
76879-210 77884-491 77884-492
6-(Hydroxymethyl)pyridine-2-carboxaldehyde 98%
6-(Hydroxymethyl)pyridine-2-carboxaldehyde

Precaución: for Research Use Only

Formula: C₇H₇NO₂
MDL Number: MFCD10700214
CAS Number: 39621-11-9
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR