3-Fluoro-4-(methylsulfonyl)benzeneboronic acid 98%

Supplier: Ambeed
Warning

Synonyms: 3-Fluoro-4-(methylsulphonyl)phenylboronic acid

A631883-1G A631883-250MG
76879-198EA 129.43 USD
76879-198 76879-200
3-Fluoro-4-(methylsulfonyl)benzeneboronic acid 98%
3-Fluoro-4-(methylsulfonyl)benzeneboronic acid

Precaución: for Research Use Only

Formula: C₇H₈BFO₄S
MW: 218.01 g/mol
MDL Number: MFCD09878323
CAS Number: 648904-83-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR