2-Bromo-4-methylthiophene 95%

Supplier: Ambeed
Danger

A631816-250MG A631816-1G A631816-5G
76879-196EA 80 USD
76879-196 77673-712 77884-489
2-Bromo-4-methylthiophene 95%
2-Bromo-4-methylthiophene

Precaución: for Research Use Only

Formula: C₅H₅BrS
MDL Number: MFCD12033287
CAS Number: 53119-60-1
UN: 1993
ADR: 3,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR