3-Methoxyphthalide 98%

Supplier: Ambeed
Warning

Synonyms: 3-Methoxyisobenzofuran-1(3H)-one, 3-Methoxy-1(3H)-isobenzofuranone

A630221-10G A630221-5G A630221-1G
76879-192EA 204.76 USD
76879-192 76879-194 77673-706
3-Methoxyphthalide 98%
3-Methoxyphthalide

Precaución: for Research Use Only

Formula: C₉H₈O₃
MW: 164.1580 g/mol
MDL Number: MFCD01652196
CAS Number: 4122-57-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR