1-Butyl-3-methylimidazoliumbis(trifluoromethylsulphonyl)imide 95%

Supplier: Ambeed
Danger

Synonyms: 1-Butyl-3-methylimidazolium Bis(trifluoromethanesulphonyl)imide, 1-Butyl-3-methyl-1H-imidazol-3-ium bis((trifluoromethyl)sulphonyl)amide

A629852-1G A629852-25G A629852-5G
76879-186EA 9 USD
76879-186 76879-188 76879-190
1-Butyl-3-methylimidazoliumbis(trifluoromethylsulphonyl)imide 95%
1-Butyl-3-methylimidazoliumbis(trifluoromethylsulphonyl)imide

Precaución: for Research Use Only

Formula: C₁₀H₁₅F₆N₃O₄S₂
MDL Number: MFCD05664714
CAS Number: 174899-83-3
UN: 2922
ADR: 8,6.1,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR