7-Quinolineboronic acid 98%

Supplier: Ambeed
Warning

Synonyms: Quinoline-7-boronic acid, 7-Quinolinylboronic acid

A628443-100MG A628443-1G A628443-250MG A628443-5G
76879-178EA 36.08 USD
76879-178 76879-180 76879-182 76879-184
7-Quinolineboronic acid 98%
7-Quinolineboronic acid

Precaución: for Research Use Only

Formula: C₉H₈BNO₂
MW: 172.9800 g/mol
MDL Number: MFCD11100750
CAS Number: 629644-82-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR