(3R,4R,5S)-Ethyl 4-(N-(tert-butyl)acetamido)-5-(diallylamino)-3-(pentan-3-yloxy)cyclohex-1-enecarboxylate hydrochloride 95% (contains < 10% EtOH)

Supplier: Ambeed
Warning

A628276-1G A628276-25G A628276-5G
76879-166EA 15.92 USD
76879-166 76879-168 76879-170
(3R,4R,5S)-Ethyl 4-(N-(tert-butyl)acetamido)-5-(diallylamino)-3-(pentan-3-yloxy)cyclohex-1-enecarboxylate hydrochloride 95% (contains < 10% EtOH)
(3R,4R,5S)-Ethyl 4-(N-(tert-butyl)acetamido)-5-(diallylamino)-3-(pentan-3-yloxy)cyclohex-1-enecarboxylate hydrochloride

Precaución: for Research Use Only

Formula: C₂₆H₄₅ClN₂O₄
MW: 485.09 g/mol
MDL Number: MFCD09032929
CAS Number: 651324-08-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR