2,4-Dihydroxy-6-methylbenzaldehyde 98%

Supplier: Ambeed
Warning

A623852-25G A623852-5G A623852-250MG
76879-140EA 542.12 USD
76879-140 77573-024 77673-686
2,4-Dihydroxy-6-methylbenzaldehyde 98%
2,4-Dihydroxy-6-methylbenzaldehyde

Precaución: for Research Use Only

Formula: C₈H₈O₃
MW: 152.147 g/mol
MDL Number: MFCD00033852
CAS Number: 487-69-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR