2-Methyl-4-(piperidin-1-ylsulfonyl)phenylboronic acid 96%

Supplier: Ambeed
Warning

Synonyms: (2-Methyl-4-(piperidin-1-ylsulphonyl)phenyl)boronic acid

A622026-100MG A622026-1G A622026-250MG
76879-134EA 37.14 USD
76879-134 76879-136 76879-138
2-Methyl-4-(piperidin-1-ylsulfonyl)phenylboronic acid 96%
2-Methyl-4-(piperidin-1-ylsulphonyl)phenylboronic acid

Precaución: for Research Use Only

Formula: C₁₂H₁₈BNO₄S
MW: 283.1500 g/mol
MDL Number: MFCD13195672
CAS Number: 1217501-52-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR