Imidazo[1,2-b]pyridazine-3-carboxylic acid 95%

Supplier: Ambeed
Warning

A620727-1G A620727-100MG
76879-120EA 353 USD
76879-120 77886-495
Imidazo[1,2-b]pyridazine-3-carboxylic acid 95%
Imidazo[1,2-b]pyridazine-3-carboxylic acid

Precaución: for Research Use Only

Formula: C₇H₅N₃O₂
MDL Number: MFCD18917066
CAS Number: 1308384-58-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR