4-[2-(Phthalimid-1-yl)ethoxy]benzeneboronic acid, pinacol ester 96%

Supplier: Ambeed
Warning

Synonyms: 2-(2-(4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenoxy)ethyl)isoindoline-1,3-dione, 2-{2-[4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenoxy]ethyl}isoindoline-1,3-dione

A620437-100MG A620437-1G A620437-250MG
76879-114EA 37 USD
76879-114 76879-116 76879-118
4-[2-(Phthalimid-1-yl)ethoxy]benzeneboronic acid, pinacol ester 96%
4-[2-(Phthalimid-1-yl)ethoxy]benzeneboronic acid, pinacol ester

Precaución: for Research Use Only

Formula: C₂₂H₂₄BNO₅
MW: 393.241 g/mol
MDL Number: MFCD09258656
CAS Number: 957061-09-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR