5-Bromo-2,3-difluorobenzeneboronic acid, pinacol ester 96%

Supplier: Ambeed
Warning

Synonyms: 2-(5-Bromo-2,3-difluorophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

A618915-1G A618915-250MG A618915-5G
76879-108EA 181.42 USD
76879-108 76879-110 76879-112
5-Bromo-2,3-difluorobenzeneboronic acid, pinacol ester 96%
5-Bromo-2,3-difluorobenzeneboronic acid, pinacol ester

Precaución: for Research Use Only

Formula: C₁₂H₁₄BBrF₂O₂
MW: 318.95 g/mol
MDL Number: MFCD11855989
CAS Number: 1073339-12-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR