Methyl 1-((tert-butoxycarbonyl)amino)cyclopropanecarboxylate 98%

Supplier: Ambeed
Warning

A618435-1G A618435-5G
76879-104EA 26.53 USD
76879-104 76879-106
Methyl 1-((tert-butoxycarbonyl)amino)cyclopropanecarboxylate 98%
Methyl 1-((tert-butoxycarbonyl)amino)cyclopropanecarboxylate

Precaución: for Research Use Only

Formula: C₁₀H₁₇NO₄
MW: 215.24 g/mol
MDL Number: MFCD11520021
CAS Number: 66494-26-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR