3-Bromo-6-piperazino-2-picoline hydrochloride 98%

Supplier: Ambeed
Warning

Synonyms: 1-(5-Bromo-6-methylpyridin-2-yl)piperazine hydrochloride

A618103-100MG A618103-1G A618103-250MG
76879-098EA 15.92 USD
76879-098 76879-100 76879-102
3-Bromo-6-piperazino-2-picoline hydrochloride 98%
3-Bromo-6-piperazino-2-picoline hydrochloride

Precaución: for Research Use Only

Formula: C₁₀H₁₅BrClN₃
MW: 292.6000 g/mol
MDL Number: MFCD12913993
CAS Number: 1187386-04-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR