2-Amino-5-chlorophenylboronic acid pinacol ester 96%

Supplier: Ambeed
Warning

Synonyms: 2-Amino-5-chlorophenyl boronic acid pinacol ester, 4-Chloro-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline

A617654-100MG A617654-1G A617654-250MG A617654-5G A617654-10G A617654-25G
76879-090EA 5 USD
76879-090 76879-092 76879-094 76879-096 77884-477 77884-478
2-Amino-5-chlorophenylboronic acid pinacol ester 96%
2-Amino-5-chlorophenylboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₂H₁₇BClNO₂
MDL Number: MFCD06795672
CAS Number: 1073371-77-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR