3-Chloro-4-cyanobenzeneboronic acid 98%

Supplier: Ambeed
Warning

Synonyms: (3-Chloro-4-cyanophenyl)boronic acid

A617240-1G A617240-250MG A617240-5G
76879-084EA 111.4 USD
76879-084 76879-086 76879-088
3-Chloro-4-cyanobenzeneboronic acid 98%
3-Chloro-4-cyanobenzeneboronic acid

Precaución: for Research Use Only

Formula: C₇H₅BClNO₂
MW: 181.382 g/mol
MDL Number: MFCD11504841
CAS Number: 1008415-02-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR