tert-Butyl 4-(1-aminoethyl)piperidine-1-carboxylate 96%

Supplier: Ambeed
Warning

A616453-100MG A616453-1G
76879-080EA 48.81 USD
76879-080 76879-082
tert-Butyl 4-(1-aminoethyl)piperidine-1-carboxylate 96%
tert-Butyl 4-(1-aminoethyl)piperidine-1-carboxylate

Precaución: for Research Use Only

Formula: C₁₂H₂₄N₂O₂
MW: 228.33 g/mol
MDL Number: MFCD11045798
CAS Number: 455267-29-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR