5-Borono-1-naphthoic acid 95%

Supplier: Ambeed
Warning

A613224-100MG A613224-1G A613224-250MG A613224-5G
76879-050EA 141.1 USD
76879-050 76879-052 76879-054 76879-056
5-Borono-1-naphthoic acid 95%
5-Borono-1-naphthoic acid

Precaución: for Research Use Only

Formula: C₁₁H₉BO₄
MW: 216 g/mol
MDL Number: MFCD09837640
CAS Number: 216060-11-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR