4-Chlorothiophene-2-carbonitrile 98%

Supplier: Ambeed
Warning

A611723-100MG A611723-1G A611723-250MG
76879-044EA 21 USD
76879-044 76879-046 76879-048
4-Chlorothiophene-2-carbonitrile 98%
4-Chlorothiophene-2-carbonitrile

Precaución: for Research Use Only

Formula: C₅H₂ClNS
MDL Number: MFCD18070966
CAS Number: 910553-55-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR