1-(4-Amino-3-fluorophenyl)ethanone 98%

Supplier: Ambeed
Warning

A609836-10G A609836-1G A609836-250MG A609836-25G A609836-5G
76879-034EA 90.18 USD
76879-034 76879-036 76879-038 76879-040 76879-042
1-(4-Amino-3-fluorophenyl)ethanone 98%
1-(4-Amino-3-fluorophenyl)ethanone

Precaución: for Research Use Only

Formula: C₈H₈FNO
MW: 153.154 g/mol
MDL Number: MFCD12032124
CAS Number: 73792-22-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR