4-Thiouracil 98%

Supplier: Ambeed
Warning

Synonyms: 4-Thioxo-3,4-dihydropyrimidin-2(1H)-one

A609629-250MG A609629-1G
76879-032EA 28.65 USD
76879-032 77613-894
4-Thiouracil 98%
4-Thiouracil

Precaución: for Research Use Only

Formula: C₄H₄N₂OS
MW: 128.15 g/mol
MDL Number: MFCD00090842
CAS Number: 591-28-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR